Skip to Content
Merck
All Photos(2)

Key Documents

A1632

Sigma-Aldrich

DL-7-Azatryptophan hydrate

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3O2 · H2O
CAS Number:
Molecular Weight:
223.23
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-7-Azatryptophan hydrate,

Assay

≥98% (TLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

O.NC(Cc1c[nH]c2ncccc12)C(O)=O

InChI

1S/C10H11N3O2.H2O/c11-8(10(14)15)4-6-5-13-9-7(6)2-1-3-12-9;/h1-3,5,8H,4,11H2,(H,12,13)(H,14,15);1H2

InChI key

PXDRHYQAIUZKHN-UHFFFAOYSA-N

Biochem/physiol Actions

DL-7-Azatryptophan is a racemic mixture of D- and L-7-azatryptophan which together with L-tryptophan is a synergistic inducer of tryptophan oxygenase of Pseudomonas acidovorans. DL-7-Azatryptophan inhibits photosynthetic carbon assimilation, photosynthetic oxygen evolution and nitrogen metabolism in Anabaena sp. Strain 1F, a marine filamentous, heterocystous cyanobacterium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C H Chen et al.
Journal of bacteriology, 169(3), 1107-1113 (1987-03-01)
The addition of DL-7-azatryptophan (AZAT), a tryptophan analog, to continuous cultures of Anabaena sp. strain CA grown with 10 mM nitrate as the nitrogen source resulted in the differentiation of heterocysts. Analysis of the intracellular amino acid pools of Anabaena
Fernando Formaggio et al.
Advances in experimental medicine and biology, 527, 731-737 (2004-06-23)
Aal and 7-Atrp, quasi-isosteric with Trp, have been inserted together with a TOAC residue in two 3(10)-helical, model hexapeptides. The interaction of photoexcited AA1 and 7-Atrp with the nitroxide group of TOAC was investigated by time resolved EPR. Both peptides
H Rosenfeld et al.
Journal of bacteriology, 97(2), 697-704 (1969-02-01)
The process of induction of tryptophan oxygenase in Pseudomonas acidovorans is typical of many microbial enzyme induction systems, in that it (i) requires cell multiplication and de novo protein synthesis, (ii) is subject to catabolite repression, (iii) results in the
P Soumillion et al.
Protein engineering, 8(5), 451-456 (1995-05-01)
The phage lambda lysozyme (lambda L) contains four tryptophans. These have been efficiently replaced by 7-azatryptophan (7aW) through biosynthetic incorporation into the overexpressed protein. Comparative analysis of the effect of temperature or pH on the fluorescence of the wild-type lambda
P Cioni et al.
Biochemical and biophysical research communications, 248(2), 347-351 (1998-07-24)
The tryptophan analogues 7-azaindole (7-Aza W) and 5-hydroxytryptophan (5-OH W) have a significant absorbance between 310-320 nm, which allows them to act as selective luminescence probes in protein mixtures containing a large number of tryptophan residues. To assess the potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service