Skip to Content
Merck
All Photos(1)

Key Documents

A1312

Sigma-Aldrich

Alternariol from Alternaria sp.

~96%

Synonym(s):

3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€728.00

€728.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€728.00

About This Item

Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€728.00


Check Cart for Availability

Request a Bulk Order

Assay

~96%

Quality Level

form

powder

storage temp.

2-8°C

SMILES string

Cc1cc(O)cc2OC(=O)c3c(O)cc(O)cc3-c12

InChI

1S/C14H10O5/c1-6-2-7(15)5-11-12(6)9-3-8(16)4-10(17)13(9)14(18)19-11/h2-5,15-17H,1H3

InChI key

CEBXXEKPIIDJHL-UHFFFAOYSA-N

General description

Alternariol belongs to the dibenzo-pyrones chemical group.[1] It is a mycotoxin present in indoor air, soil and plants.[2]

Application

Alternariol from Alternaria sp. may be used as a calibration standard in reverse phase high-performance liquid chromatography (HPLC) and UV-spectrum analysis.[2] It may also be used to spike wholemeal wheat flour for wet baking studies [3]

Biochem/physiol Actions

Alternaria mycotoxins are generally associated with undried food grains post-harvest.[2] The disease caused by Alternaria mycotoxin results in a discolored halo in fruits and spotting in leaves.[2] Alternariol (AOH) is cytotoxic and fetotoxic to microbes and mammalian cells.[2] AOH displays inhibitory effect on cell proliferation and has estrogenic functionality.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Siegel et al.
Journal of agricultural and food chemistry, 58(17), 9622-9630 (2010-08-07)
The stability of the Alternaria mycotoxins alternariol, alternariol monomethyl ether, and altenuene upon bread baking was investigated by model experiments using a spiked wholemeal wheat flour matrix. For alternariol and alternariol monomethyl ether, but not for altenuene, degradation products, formed
Alternaria mycotoxins in food and feed: an overview
EscrivaL, et al.
Journal of Food Quality, 2017 (2017)
Eva Graf et al.
International journal of food microbiology, 157(3), 353-359 (2012-06-26)
Strains of the genus Alternaria are ubiquitously present and frequently found on fruits, vegetables and cereals. One of the most commonly found species from this genus is A. alternata which is able to produce the mycotoxin alternariol among others. To
A Solhaug et al.
Mutation research, 738-739, 1-11 (2012-10-04)
Alternariol (AOH), a mycotoxin produced by Alternaria sp, is often found as a contaminant in fruit and cereal products. Here we employed the murine macrophage cell line RAW 264.7 to test the hypothesis that AOH causes toxicity as a response
Yvonne Ackermann et al.
Journal of agricultural and food chemistry, 59(12), 6360-6368 (2011-05-04)
This study investigated the production of polyclonal (pAB) antibodies and the first time production of monoclonal (mAB) antibodies against the mycotoxin alternariol, and their implementation in enzyme immunoassay (EIA) for the rapid determination of alternariol in foods. Both EIAs were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service