All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H13NO4
CAS Number:
Molecular Weight:
223.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
storage temp.
−20°C
SMILES string
N[C@H](CC(O)=O)C(=O)OCc1ccccc1
InChI
1S/C11H13NO4/c12-9(6-10(13)14)11(15)16-7-8-4-2-1-3-5-8/h1-5,9H,6-7,12H2,(H,13,14)/t9-/m1/s1
InChI key
NJSRYBIBUXBNSW-SECBINFHSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amit Kumar et al.
Frontiers in molecular neuroscience, 11, 327-327 (2018-10-03)
Two major questions in the field of Alzheimer-type dementia remain elusive. One is the native function of amyloid-β (Aβ) peptides and the other is an early deficit in the central cholinergic network. Nevertheless, recent evidence suggests that Aβ peptides are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service