Skip to Content
Merck
All Photos(1)

Documents

A0936

Sigma-Aldrich

N-Acetylmuramyl-Ala-D-isoglutaminyl-Nε-stearoyl-Lys

Synonym(s):

MDP-LYS(L18), Muroctasin, Romurtide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H78N6O13
CAS Number:
Molecular Weight:
887.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)NCCCC[C@H](NC(=O)CC[C@@H](NC(=O)[C@H](C)NC(=O)[C@@H](C)O[C@H]1[C@H](O)[C@@H](CO)OC(O)[C@@H]1NC(C)=O)C(N)=O)C(O)=O

InChI

1S/C43H78N6O13/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23-34(52)45-26-21-20-22-32(42(58)59)48-35(53)25-24-31(39(44)55)49-40(56)28(2)46-41(57)29(3)61-38-36(47-30(4)51)43(60)62-33(27-50)37(38)54/h28-29,31-33,36-38,43,50,54,60H,5-27H2,1-4H3,(H2,44,55)(H,45,52)(H,46,57)(H,47,51)(H,48,53)(H,49,56)(H,58,59)/t28-,29+,31+,32-,33+,36+,37+,38+,43?/m0/s1

InChI key

FKHUGQZRBPETJR-FZKCHLSLSA-N

Biochem/physiol Actions

MDP-LYS(L18), a synthetic analogue of MDP (muramyl dipeptide) derived from bacterial cell walls, is used as a potent immunoadjuvant that enhances protective immunity through activation of monocyte-macrophage lineage cells, such as dendritic cells.
N-Acetylmuramyl-Ala-D-isoglutaminyl-Nε-stearoyl-Lys, Romurtide, a muramyl dipeptide analogue, is a granulocyte-macrophage colony-stimulating factor and a TNF-α inducer.
Antibiotic peptide.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fukushima et al.
Vaccine, 14(6), 485-491 (1996-04-01)
To examine the effect of MDP-Lys(L18), a derivative of muramyl dipeptide (MDP), as a mucosal immunoadjuvant, we investigated its activity to augment host resistance against mucosal infections by Sendai virus and rotavirus in mice. In an experimental infection model using
H Ueda et al.
International immunopharmacology, 1(1), 97-104 (2001-05-23)
The novel applications of a muramyl dipeptide analogue, romurtide, were investigated. When mice were intravenously injected with 10-1000 micrograms/mouse of romurtide as a primer and 3 KE/mouse of OK-432 as a trigger, systemic tumor necrosis factor-alpha (TNF-alpha) activity was increased.
T Sugawara et al.
International journal of tissue reactions, 17(1), 5-13 (1995-01-01)
To examine the involvement of lymphocytes in the development of MDP-Lys(L18)-induced arthritis (MIA) in rats and the exacerbation of MIA by cyclosporin A (CsA), we analysed the spleen lymphocyte subset using monoclonal antibodies and flow cytometry during the development of
K Namba et al.
Vaccine, 15(4), 405-413 (1997-03-01)
The response of megakaryocytes and cytokines to the administration of romurtide, a synthetic muramyl dipeptide derivative, was investigated in monkeys with myelosuppression by carboplatin-treatment. Romurtide increased the number of megakaryocytes and promoted the shift of megakaryocytes towards high ploidy class
T Sugawara et al.
Inflammation, 20(1), 43-56 (1996-02-01)
To examine the involvement of cytokines in the mechanisms of N2-[(N-acetylmuramoyl)-L-alanyl-D-isoglutaminyl]-N6-stearoyl-L-lysine, MDP-Lys(L18)-induced arthritis, we analyzed interleukin-1 (IL-1), tumor necrosis factor (TNF), colony-stimulating factor (CSF), and neutrophil chemotactic factor (NCF) by bioassays in the rat macrophage-conditioned medium (Mluminal diameter-CM) stimulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service