Skip to Content
Merck

A0724

N-Acetylallolactosamine

≥90%, solid

Synonym(s):

β-galactosyl-(1-6)-N-acetylglucosamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H25NO11
Molecular Weight:
383.35
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90%

form

solid

storage temp.

2-8°C

SMILES string

CC(=O)NC(C=O)C(O)C(O)C(O)COC1OC(CO)C(O)C(O)C1O

InChI

1S/C14H25NO11/c1-5(18)15-6(2-16)9(20)10(21)7(19)4-25-14-13(24)12(23)11(22)8(3-17)26-14/h2,6-14,17,19-24H,3-4H2,1H3,(H,15,18)

InChI key

FGYNENQLWILFLQ-UHFFFAOYSA-N

Application

N-Acetylallolactosamine is used to study and differentiate hemagglutination mediators such as the L10b lectin from horseshoe crab hemocytes.

Biochem/physiol Actions

Starting material for carbohydrate syntheses

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library