A0679
3′-Azido-3′-deoxythymidine β-D-glucuronide sodium salt
≥97% (HPLC)
Synonym(s):
AZT glucuronide sodium-potassium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C16H20N5O10Na
CAS Number:
Molecular Weight:
465.35
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
≥97% (HPLC)
Other Notes
Glucuronide metabolite of 3′-azido-3′-deoxythymidine
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Okuda et al.
Journal of chromatography. B, Biomedical sciences and applications, 726(1-2), 225-236 (1999-05-29)
A complete two-dimensional separation technique for the determination of acidic compounds in plasma was developed by using column-switching reversed-phase liquid chromatography. This technique was based on solute peak enrichment at the top of the second column during heart-cutting and an
M Garland et al.
Pediatric research, 44(1), 47-53 (1998-07-17)
Zidovudine (azidothymidine, AZT) is used in pregnancy to reduce mother to infant transmission of HIV. Understanding the disposition of AZT in the fetus is necessary to optimize therapeutic regimens directed toward the fetus. Recent studies in primates found similar steady-state
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service