Skip to Content
Merck

A0591

Autocamtide 2 trifluoroacetate salt

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C65H118N22O20
CAS Number:
Molecular Weight:
1527.77
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥60%

color

white

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](C)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCCCN)[C@@H](C)O)C(C)C)C(O)=O

InChI

1S/C65H118N22O20/c1-31(2)28-43(83-51(94)34(7)76-54(97)38(17-11-13-25-67)78-53(96)37(68)16-10-12-24-66)60(103)80-40(19-15-27-75-65(72)73)55(98)79-39(18-14-26-74-64(70)71)56(99)81-41(20-22-46(69)89)57(100)82-42(21-23-47(90)91)58(101)87-50(36(9)88)62(105)86-49(33(5)6)61(104)84-44(30-48(92)93)59(102)77-35(8)52(95)85-45(63(106)107)29-32(3)4/h31-45,49-50,88H,10-30,66-68H2,1-9H3,(H2,69,89)(H,76,97)(H,77,102)(H,78,96)(H,79,98)(H,80,103)(H,81,99)(H,82,100)(H,83,94)(H,84,104)(H,85,95)(H,86,105)(H,87,101)(H,90,91)(H,92,93)(H,106,107)(H4,70,71,74)(H4,72,73,75)/t34-,35-,36+,37-,38-,39-,40-,41-,42-,43-,44-,45-,49-,50-/m0/s1

InChI key

WRSMVHZKPDCKNQ-DBSTUJSUSA-N

Gene Information

human ... CAMK1(8536)
mouse ... CAMK1(52163)
rat ... CAMK1(171503)

General description

A highly selective synthetic Ca2+/calmodulin-dependent protein kinase substrate. The peptide contains the sequence around the autophosphorylation site in the autoinhibitory domain of calmodulin-dependent protein kinase II (CaM kinase II).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library