Skip to Content
Merck

A0577

Aluminum 1,8,15,22-tetraphenoxy-29H,31H-phthalocyanine chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C56H32AlClN8O4
CAS Number:
Molecular Weight:
943.34
UNSPSC Code:
12171500
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

Dye content, ~80%

SMILES string

Cl[Al]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7c(Oc8ccccc8)cccc37)c9cccc(Oc%10ccccc%10)c69)c%11cccc(Oc%12ccccc%12)c5%11)c%13cccc(Oc%14ccccc%14)c4%13



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library