Skip to Content
Merck

Skip To

A0384

Arcaine sulfate salt

Synonym(s):

1,4-Diguanidinobutane sulfate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 108,00

About This Item

Empirical Formula (Hill Notation):
C6H16N6 · H2O4S
CAS Number:
Molecular Weight:
270.31
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (TLC)
Form:
powder

€ 108,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥98% (TLC)

form

powder

mp

>280 °C

solubility

1 M NaOH: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OS(O)(=O)=O.NC(=N)NCCCCNC(N)=N

InChI

1S/C6H16N6.H2O4S/c7-5(8)11-3-1-2-4-12-6(9)10;1-5(2,3)4/h1-4H2,(H4,7,8,11)(H4,9,10,12);(H2,1,2,3,4)

InChI key

RWTGFMPOODRXIM-UHFFFAOYSA-N

Application

Arcaine sulfate salt has been used as an N-methyl-d-aspartate (NMDA) receptor effector/antagonist:
  • to evaluate its effects on the acquisition of electrical kindling of the amygdala[1]
  • to compare its effects with agmatine and MK801 on NMDA- or glutamate-induced neuronal damage[2]
  • to study its effects on cell-free translation of synthetic polynucleotide[3]

Biochem/physiol Actions

Arcaine sulfate salt (1,4-diguanidinobutane), a polyamine,[3] is an antagonist of the polyamine binding site of the N-methyl-d-aspartate (NMDA) receptor.[4] Arcaine, an agmatine analog,[2] along with ifenprodil, might have a possibility for therapeutic management in epileptic disorders.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M7033A7127H7278
assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥97%

assay

≥98% (TLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

solubility

1 M NaOH: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

solubility

water: 49.00-51.00 mg/mL, clear to very slightly hazy, colorless to faintly yellow

solubility

H2O: 50 mg/mL, ethanol: insoluble

solubility

acetic acid: 10 mg/mL, clear, colorless to faintly yellow

mp

>280 °C

mp

-

mp

234-238 °C (lit.)

mp

204 °C

Gene Information

human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRINA(2907)

Gene Information

human ... NOS1(4842), NOS2(4843), NOS3(4846)
mouse ... Nos2(18126)
rat ... Nos1(24598), Nos2(24599)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152), GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRINA(2907)

Gene Information

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters