Skip to Content
Merck
All Photos(1)

Key Documents

A0184

Sigma-Aldrich

Amyloid β-Protein Fragment 1-28

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C145H209N41O46
CAS Number:
Molecular Weight:
3262.46
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc5ccccc5)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](Cc6ccccc6)C(=O)N[C@@H](Cc7ccccc7)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](C(C)C)C(=O)NCC(=O)N[C@@H](CO)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCCN)C(O)=O

Gene Information

human ... APP(351)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys

Biochem/physiol Actions

β-Amyloid peptide that forms fibrils in vitro that have the same structure and antigenicity as those found in Alzheimer′s patients.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X A Alvarez et al.
Methods and findings in experimental and clinical pharmacology, 19(7), 471-479 (1997-12-31)
beta-amyloid (beta A) deposition is a key event in the etiopathogenesis of Alzheimer's disease (AD), contributing to neuronal degeneration and cognitive impairment in AD patients. Both neurotrophic and neurotoxic actions of beta A have been demonstrated in experimental conditions. In
T G Fletcher et al.
Protein science : a publication of the Protein Society, 6(3), 666-675 (1997-03-01)
The neurotoxicity of beta-amyloid protein (beta AP) fragments may be a result of their solution conformation, which is very sensitive to solution conditions. In this work we describe NMR and CD studies of the conformation of beta AP(12-28) in lipid
J J Miguel-Hidalgo et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 8(3), 203-208 (1998-08-26)
We assessed the neuroprotective capabilities of S12024 (R,S 1-methyl 8-(2-morpholinylmethoxy)-1,2,3,4-tetrahydroquinoleine methane sulphonate) in a model of neuronal degeneration in the dentate gyrus of the rat hippocampus. Specific degeneration of a large part of neurons in the lateral blade of the
K Kirshenbaum et al.
Biochemistry, 34(23), 7629-7639 (1995-06-13)
Molecular dynamics simulations were used to successfully reproduce the experimentally observed pH-dependent conformational behavior of a monomeric peptide in aqueous solution. Simulations were conducted at 298 K on a peptide corresponding to residues 1-28 of the amyloid beta-peptide [referred to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service