Skip to Content
Merck

A002

N6-Cyclohexyladenosine

Synonym(s):

CHA

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C16H23N5O4
CAS Number:
Molecular Weight:
349.38
UNSPSC Code:
12352204
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(NC4CCCCC4)ncnc23

InChI

1S/C16H23N5O4/c22-6-10-12(23)13(24)16(25-10)21-8-19-11-14(17-7-18-15(11)21)20-9-4-2-1-3-5-9/h7-10,12-13,16,22-24H,1-6H2,(H,17,18,20)/t10-,12-,13-,16-/m1/s1

InChI key

SZBULDQSDUXAPJ-XNIJJKJLSA-N

General description

N6-Cyclohexyladenosine belongs to the class of new generation thermolytic agents.

Biochem/physiol Actions

N6-Cyclohexyladenosine is an adenosine A1 receptor agonist. It prevents thermogenesis and other response such as shivering in response to cold. N6-Cyclohexyladenosine initiates hibernation in animals.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of conventional outflow facility by the adenosine A1 agonist N6-cyclohexyladenosine
Crosson CE, et al.
Investigative Ophthalmology & Visual Science, 46(10), 3795-3799 (2005)
Precise Control of Target Temperature Using N6-Cyclohexyladenosine and Real-Time Control of Surface Temperature
Laughlin BW, et al.
Therapeutic hypothermia and temperature management, 8(2), 108-116 (2018)
Matthew Edward Pamenter et al.
Brain research, 1213, 27-34 (2008-05-06)
Adenosine is a defensive metabolite that is critical to anoxic neuronal survival in the freshwater turtle. Channel arrest of the N-methyl-d-aspartate receptor (NMDAR) is a hallmark of the turtle's remarkable anoxia tolerance and adenosine A1 receptor (A1R)-mediated depression of normoxic
Gerard J Marek
Neuropharmacology, 56(8), 1082-1087 (2009-03-28)
Modulation of glutamatergic neurotransmission by metabotropic glutamate2/3 (mGlu2/3) receptor agonists effectively treats seemingly diverse neuropsychiatric illness such as generalized anxiety disorder and schizophrenia. Activation of adenosine A(1) heteroceptors, like mGlu2 autoreceptors, decreases glutamate release in the medial prefrontal cortex (mPFC)
Kelly R Lobato et al.
Progress in neuro-psychopharmacology & biological psychiatry, 32(4), 994-999 (2008-02-22)
It was previously shown that the acute administration of zinc chloride elicits an antidepressant-like effect in the mouse forced swimming test (FST). We have also shown that the activation of adenosine A(1) and A(2A) receptors produces an antidepressant-like effect in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service