Skip to Content
Merck
All Photos(1)

Key Documents

P0115

Sigma-Aldrich

PAC-1

≥98% (HPLC)

Synonym(s):

(4-Benzylpiperazino)acetic acid (3-allyl-2-hyroxybenzylidene)hydrazide, 1-Piperazineacetic acid, 2-(phenylmethyl)-,[[2-hydroxy-3-(2-propenyl)phenyl]methylene]hyrrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€364.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€364.00

About This Item

Empirical Formula (Hill Notation):
C23H28N4O2
CAS Number:
Molecular Weight:
392.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€112.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: ≥10 mg/mL
deionized water: ≤2 mg/mL

storage temp.

room temp

SMILES string

Oc1c(CC=C)cccc1\C=N\NC(=O)CN2CCN(CC2)Cc3ccccc3

InChI

1S/C23H28N4O2/c1-2-7-20-10-6-11-21(23(20)29)16-24-25-22(28)18-27-14-12-26(13-15-27)17-19-8-4-3-5-9-19/h2-6,8-11,16,29H,1,7,12-15,17-18H2,(H,25,28)/b24-16+

InChI key

YQNRVGJCPCNMKT-LFVJCYFKSA-N

General description

Procaspase-activating compound 1 (PAC-1) is an ortho-hydroxy N-acyl hydrazine compound.[1] It is cytotoxic to a variety of cancer cells, such as lymphoma, leukaemia, breast carcinoma and glioblastoma.[2] High doses of PAC-1 can promote neuroexcitation.[2] It helps in the in vitro autoactivation of the proenzyme protease, which acts as a prototype for other protease activating compounds.[3] PAC-1 dependant activation of procaspase-3 is associated with the chelation of zinc.[1]

Biochem/physiol Actions

PAC-1 is a caspase 3 activator.
PAC-1 is a caspase 3 activator. Caspase activation is a key strategy in cancer therapy. Caspase 3 is a key executioner caspase and direct effector of apoptosis. Hallmark of many cancers is the circumvention of signal in the activation of executioner caspases and loss of apoptotic response. EC50 for caspase 3 activation = 0.33 μM and caspase 7= 4.5 μM. The IC50 for apoptosis induction = 0.92 μM. Elevated caspase 3 level in cancerous cell lines and tissues allows PAC-1 to selectively induce apoptosis in tissues.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Philipp Blüm et al.
Cellular & molecular immunology, 14(12), 986-996 (2016-04-19)
Hepatitis C virus (HCV) infection is a major problem worldwide. HCV is not limited to liver disease but is frequently complicated by immune-mediated extrahepatic manifestations such as glomerulonephritis or vasculitis. A fatal complication of HCV-associated vascular disease is thrombosis. Polyriboinosinic:polyribocytidylic
Targeting of C-type lectin-like receptor 2 or P2Y12 for the prevention of platelet activation by immunotherapeutic CpG oligodeoxynucleotides: comment.
U Flierl et al.
Journal of thrombosis and haemostasis : JTH, 16(1), 181-185 (2017-10-21)
Michal Bijak et al.
International journal of biological macromolecules, 106, 878-884 (2017-08-27)
The primary biological function of platelets is to form hemostatic thrombi that prevent blood loss and maintain vascular integrity. These multi-responding cells are activated by different endogenous, physiological agonists due to the vast number of receptors present on the surface
Parallel synthesis and biological evaluation of 837 analogues of procaspase-activating compound 1 (PAC-1)
Hsu DC et al.
ACS Combinatorial Science, 14(1), 44-50 (2011)
Procaspase-3 activation as an anti-cancer strategy: structure- activity relationship of procaspase-activating compound 1 (PAC-1) and Its cellular co-localization with caspase-3
Peterson QP et al.
Journal of Medicinal Chemistry, 52(18), 5721-5731 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service