Skip to Content
Merck

957D

DAB Substrate Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12161503
Technique(s):
immunohistochemistry: suitable
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For In Vitro Diagnostic Use in Select Regions

Quality Segment

form

aqueous solution

packaging

pkg of 1000 tests (957D-60), pkg of 150 tests (957D-50), pkg of 2000 tests (957D-30), pkg of 500 tests (957D-20), pkg of 5000 tests (957D-40)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable

storage temp.

2-8°C

General description

DAB Substrate Kit is intended for use in immunohistochemistry (IHC) staining protocols. This reagent is designed to detect target antigens in formalin-fixed, paraffin-embedded tissue sections when used in conjunction with primary antibodies and appropriate secondary antibody conjugates in the IHC staining process.When in the presence of HRP polymer, DAB produces a brown precipitate that can be readily visualized using light microscopy. Kits contain: DAB chromogen and DAB buffer substrate.

Physical form

Ready to use reagent

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Analysis Note

United States - IVD
Canada - IVD
European Union - IVD
Japan - RUO

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - Muta. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library