Skip to Content
Merck

Skip To

95255

D-α-Tocopherol succinate

BioXtra, ≥98.0% (HPLC)

Synonym(s):

Vitamin E succinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 208,00

About This Item

Empirical Formula (Hill Notation):
C33H54O5
CAS Number:
Molecular Weight:
530.78
UNSPSC Code:
12352211
NACRES:
NA.79
PubChem Substance ID:
EC Number:
224-403-8
Beilstein/REAXYS Number:
4038233
MDL number:
eCl@ss:
34058016

€ 208,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

assay

≥98.0% (HPLC)

form

powder

color

white to faint yellow

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(O)=O)c(C)c(C)c2O1

InChI

1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1

InChI key

IELOKBJPULMYRW-NJQVLOCASA-N

General description

Tocopherol is a lipid soluble vitamin E. It is present in food sources, such as soybean, almond oil, sunflower oil. Tocopherol has four isoforms in plants, which are α, β, γ and δ. It is composed of a chromanol ring with varied position of methyl group. D-αTocopherol succinate is also known as vitamin E succinate (VES).

Biochem/physiol Actions

Tocopherol exhibits antioxidant property by protecting the cells and membranes from oxidative stress. In plants, high levels of α-tocopherol leads to plant stress tolerance whereas low levels is associated with oxidative stress. In humans, tocopherol prevents cancer, premature aging, atherosclerosis, arthritis caused due to free radicals. Vitamin E succinate (VES) is an anti-tumor analog of vitamin E.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3126166780347782
assay

≥98.0% (HPLC)

assay

-

assay

-

assay

-

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

form

powder

form

powder

form

-

form

-

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

100

color

white to faint yellow

color

white to yellow

color

-

color

-

product line

BioXtra

product line

-

product line

-

product line

-


Still not finding the right product?

Explore all of our products under D-α-Tocopherol succinate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters