Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
250 g | Check Cart for Availability | € 80,60 |
About This Item
form
powder
Quality Segment
color
beige
viscosity
600-1500 mPa.s, 2 % in H2O(20 °C)
transition temp
flocculation range 60-90 °C (0.5 wt. %)
SMILES string
O3[C@H]([C@@H]([C@H]([C@@H]([C@H]3COC)OC)OC)OC)O[C@@H]4[C@H](O[C@H]([C@@H]([C@H]4OC)OC)OC)COC.O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O)CO.OCCO
InChI
1S/C20H38O11.C12H22O11.C2H6O2/c1-21-9-11-13(23-3)15(24-4)18(27-7)20(30-11)31-14-12(10-22-2)29-19(28-8)17(26-6)16(14)25-5;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17;3-1-2-4/h11-20H,9-10H2,1-8H3;3-20H,1-2H2;3-4H,1-2H2/t11-,12-,13-,14-,
InChI key
JTWZVKMWOCBJEV-RXUSYSPUSA-N
Application
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 200 |
| form powder | form powder | form powder | form powder |
| color beige | color white to faint beige | color white to off-white | color white to off-white |
| transition temp flocculation range 60-90 °C (0.5 wt. %) | transition temp flocculation range 60-90 °C (0.5 wt. %) | transition temp - | transition temp - |
| viscosity 600-1500 mPa.s, 2 % in H2O(20 °C) | viscosity 150-450 mPa.s, 2 % in H2O(20 °C) | viscosity 1200-1800 mPa.s, 2 % in H2O(20 °C) | viscosity 10-25 mPa.s, 2 % in H2O(20 °C) |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

