Skip to Content
Merck
All Photos(1)

Key Documents

93428

Sigma-Aldrich

Triton X-114 solution

ampule, ~10% in H2O

Synonym(s):

(1,1,3,3-Tetramethylbutyl)phenyl-polyethylene glycol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

quality

ampule

concentration

~10% in H2O

impurities

≤150 μM sulfhydryl oxidizing substances (carbonyls, peroxides)

refractive index

n20/D 1.349

CMC

0.2

transition temp

cloud point 23 °C

density

1.006 g/mL at 20 °C

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

HLB

12.4

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamta Shoaee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 70-75 (2012-09-18)
A cloud-point extraction process coupled to ICP-OES by using 3-nitro benzaldehyde thiosemicarbazone (3-NBT) as complexing agent was developed for the simultaneous preconcentration and determination of copper and mercury in water samples. The variables affecting the complexation and extraction steps were
Demirhan Citak et al.
Journal of AOAC International, 95(4), 1170-1175 (2012-09-14)
A cloud point extraction procedure was optimized for the separation and preconcentration of lead(II), cadmium(II), copper(II), and iron(III) ions in various water and canned food samples. The metal ions formed complexes with 2,6-diamino-4-phenyl-1,3,5-triazine that were extracted by surfactant-rich phases in
Ebru Ates et al.
Journal of AOAC International, 94(6), 1853-1862 (2012-02-11)
A cloud point extraction method is reported using LC/MS for the determination of regulated water-soluble food colors (Allura Red, Sunset Yellow, erythrosine, and tartrazine) and banned fat-soluble synthetic azo dyes (Sudan I, II, III, and IV; Red B; 7B; Black
Trícia Maria Ferreira de Sousa Oliveira et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 24(2), 220-222 (2015-07-15)
The relevance of the dog as a source of visceral leishmaniasis infection is known, but the role of cats as reservoir hosts for leishmaniasis is not yet fully clear. This study assessed the efficacy of conjunctival swab PCR (CS-PCR) in
Rui Ma et al.
Analytical biochemistry, 424(2), 124-126 (2012-03-01)
Triton X-114 temperature transition extraction has been considered to be a simple and cost-effective strategy to eliminate endotoxin from plasmid preparations. However, a repeated cooling-heating process may promote the degradation of plasmid DNA. Based on the finding that the cloud

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service