Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human kidney
Growth mode:
Adherent
Pricing and availability is not currently available.
biological source
human kidney
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
Human Renal Proximal Tubular Epithelial Cells (HRPTEpC) provide a useful tool for studying various aspects of pathology and biology of the human renal proximal tubular cells in vitro. HRPTEpC have recently been used in a study of metabolic enzymes responding to energy depletion in renal cells. HRPTEpCs have a pivotal role in kidney function including reabsorbtion of glucose and protein, and also play a part in the glucotoxicity associated with diabetes.
Human Renal Proximal Tubular Epithelial Cells (HRPTEpC) provide a useful tool for studying various aspects of pathology and biology of the human renal proximal tubular cells in vitro. HRPTEpC have recently been used in a study of metabolic enzymes responding to energy depletion in renal cells. HRPTEpCs have a pivotal role in kidney function including reabsorbtion of glucose and protein, and also play a part in the glucotoxicity associated with diabetes.
Application
Kidney disease, diabetes, glucotoxicity, pathology. kidney function, reabsorbtion of glucose and protein, biology of renal proximal tubular cells in vitro, metabolic enzymes, cell energy usage.
Biochem/physiol Actions
Kidney
Packaging
First or second passage, >500,000 cells in RenaEpi Basal Medium containing 10% FBS & 10% DMSO.
Analysis Note
Each lot was tested for proper morphology, ability to attach and spread on tissue culture ware surface, and proliferate in RenaEpi Growth Medium, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.
Other Notes
Can be cultured at least 6 population doublings
RenaEpi Basal Medium containing 10% FBS & 10% DMSO
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.