Skip to Content
Merck
All Photos(1)

Key Documents

91216

Sigma-Aldrich

(±)-Dihydrokaempferol

≥95% (HPLC)

Synonym(s):

(2RS,3RS)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-one, (±)-Aromadendrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

powder

Related Categories

Biochem/physiol Actions

Flavanonol; (+)-Aromadendrin is an intermediate in flavonoid biosynthesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service