Skip to Content
Merck
All Photos(1)

Key Documents

903H-0

Sigma-Aldrich

MART-1 (M2-7C10) + Tyrosinase (T311) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

M2-7C10 + T311, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (903H-07)
bottle of 7.0 mL predilute (903H-08)

isotype

IgG2a
IgG2bκ

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MLANA(2315)

General description

MART-1 (also known as Melan A) is a melanocyte differentiation antigen. It is present in melanocytes of normal skin and retina, nevi and in more than 85% of melanomas. Tyrosinase is an enzyme integral in the process of melanin synthesis, and found in 85% to 90% of malignant melanomas. Given these statistics, this cocktail is ideally suited to detection of melanomas and melanocytic lesions.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepali Gupta et al.
The American journal of surgical pathology, 26(11), 1450-1457 (2002-11-01)
Malignant melanomas of the vagina are rare tumors. In this study we present the clinicopathologic features and immunohistochemical analysis of 26 such cases seen in our institution over a period of 30 years. The patients' age ranged from 38 to
T J de Vries et al.
The Journal of pathology, 193(1), 13-20 (2001-02-13)
With the recent availability of novel antibodies against melanoma antigens tyrosinase and MART-1, it is important to validate their usefulness in pathology practice and in screening patients for immunotherapy treatment. In the present study conducted by the Melanoma Cooperative Group
Guy Orchard
British journal of biomedical science, 59(4), 196-202 (2003-02-08)
Incidence of malignant melanoma (MM) is rising rapidly throughout the Western world, and the number of melanocytic lesions removed for histological assessment has increased. MM can present with a myriad of histological appearances that make diagnosis problematic, particularly when dealing
Vinod B Shidham et al.
BMC cancer, 3, 15-15 (2003-05-09)
MART-1, Melan-A, and Tyrosinase have shown encouraging results for evaluation of melanoma micrometastases in sentinel lymph nodes, as compared to conventionally used S-100 protein and HMB-45. To achieve higher sensitivity, some studies recommend evaluation of three sections, each at intervals
M L Prasad et al.
The American journal of surgical pathology, 25(6), 782-787 (2001-06-08)
Malignant melanomas of the oral and sinonasal mucosa are rare tumors. Amelanotic variants can, on occasion, be difficult to recognize by routine light microscopy. Immunohistochemical studies may be needed for a final diagnosis. A number of new monoclonal antibodies to

Articles

IHC antibodies enhance dermatopathology beyond H&E stained slides, improving techniques and applications for dermatological research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service