Skip to Content
Merck

Skip To

88596

Fluorescein-O′-acetic acid

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Synonym(s):

FOAA, O′-(Carboxymethyl)fluorescein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 143,00
25 mg
Check Cart for Availability
€ 546,00
€ 382,20

About This Item

Empirical Formula (Hill Notation):
C22H14O7
CAS Number:
Molecular Weight:
390.34
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:

€ 143,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥90% (HPCE)

form

powder

fluorescence

λex 455 nm; λem 514 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

OC(=O)COc1ccc2c(OC3=CC(=O)C=CC3=C2c4ccccc4C(O)=O)c1

InChI

1S/C22H14O7/c23-12-5-7-16-18(9-12)29-19-10-13(28-11-20(24)25)6-8-17(19)21(16)14-3-1-2-4-15(14)22(26)27/h1-10H,11H2,(H,24,25)(H,26,27)

InChI key

VQKDLLIKXLDPHW-UHFFFAOYSA-N

Other Notes

Fluorescent label which reacts with amines after activation to give isomerically homogeneous derivatives

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
469434693553608
description

BioReagent, suitable for fluorescence, ≥90% (HPCE)

description

suitable for fluorescence, ≥97.0% (HPCE)

description

suitable for fluorescence, ≥90% (HPCE)

description

BioReagent, suitable for fluorescence, ≥90% (HPCE)

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

fluorescence

λex 455 nm; λem 514 nm in 0.1 M Tris pH 8.0

fluorescence

λex 490 nm; λem 514 nm in 0.1 M Tris pH 8.0 (esterase)

fluorescence

λex 491 nm; λem 515 nm in 0.1 M Tris pH 9.0

fluorescence

λex 490 nm; λem 524 nm in 0.1 M Tris pH 8.0

form

powder

form

powder

form

solid

form

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

assay

≥90% (HPCE)

assay

≥97.0% (HPCE)

assay

≥90% (HPCE)

assay

≥90% (HPCE)

product line

BioReagent

product line

-

product line

-

product line

BioReagent


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters