Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | € 23,00 |
1 L | Check Cart for Availability | € 97,40 |
About This Item
NACRES:
NA.25
UNSPSC Code:
12161700
€ 23,00
Check Cart for Availability
grade
Molecular Biology
Quality Segment
product line
BioReagent
form
powder blend
impurities
DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected
solubility
H2O: 0.263 g/mL, clear, colorless
storage temp.
room temp
General description
SSC Buffer powder is a standard reagent in Southern and Northern blotting procedures. This powder can be used to make SSC working solutions with concentrations ranging from 0.5x to 20x.
Application
Suitable for:
- Preparing nucleic acids for hybridization
- Hybridization buffer for Northern and Southern blots
- Washing buffer for Northern and Southern blots
Preparation Note
- Convenient pack sizes
- Multi-purpose use in hybridizations
- Easy-to-use powder makes fast, inexpensive solutions
Dissolve 263.5 g in a final volume of 1L molecular biology grade water. The resulting 20x concentrate is approximately pH 7, and can be further diluted with water, as needed.
Other Notes
SSC Buffer powder contains 0.3 M sodium citrate in 3M NaCl.
1 of 1
This Item | |||
|---|---|---|---|
| grade Molecular Biology | grade - | grade - | grade - |
| Quality Level 100 | Quality Level 200 | Quality Level - | Quality Level 100 |
| product line BioReagent | product line - | product line - | product line pHast Pack™ |
| form powder blend | form powder blend | form solution | form powder |
| solubility H2O: 0.263 g/mL, clear, colorless | solubility water: 263 mg/mL, clear, colorless | solubility - | solubility - |
| storage temp. room temp | storage temp. room temp | storage temp. 20-25°C | storage temp. - |
Still not finding the right product?
Explore all of our products under SSC Buffer
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



