Skip to Content
Merck
All Photos(1)

Key Documents

84071

Sigma-Aldrich

Ruthenium Red

for microscopy, ≥85% (calc. on dry substance, AT)

Synonym(s):

Ammoniated ruthenium oxychloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
CAS Number:
Molecular Weight:
786.35
Colour Index Number:
77800
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

for microscopy

Quality Level

Assay

≥85% (calc. on dry substance, AT)

impurities

~5% water

SMILES string

N.N.N.N.N.N.N.N.N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++]O[Ru++]O[Ru++]

InChI

1S/6ClH.14H3N.2O.3Ru/h6*1H;14*1H3;;;;;/q;;;;;;;;;;;;;;;;;;;;;;3*+2/p-6

InChI key

ZXYSDUPLHOJIOR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Inhibits mitochondrial Ca2+ uniporter; capsaicin antagonist; blocks VR1 vanilloid receptor-coupled ion channels.

Other Notes

Interaction with calcium-binding proteins

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Charuk et al.
Analytical biochemistry, 188(1), 123-131 (1990-07-01)
The interaction of ruthenium red, [(NH3)5Ru-O-Ru(NH3)4-O-Ru(NH3)5]Cl6.4H2O, with various Ca2(+)-binding proteins was studied. Ruthenium red inhibited Ca2+ binding to the sarcoplasmic reticulum protein, calsequestrin, immobilized on Sepharose 4B. Furthermore, ruthenium red bound to calsequestrin with high affinity (Kd = 0.7 microM;
Cyrielle Corbin et al.
Ultrasonics sonochemistry, 26, 176-185 (2015-03-11)
Flaxseed accumulates in its seedcoat a macromolecular complex composed of lignan (secoisolariciresinol diglucoside, SDG), flavonol (herbacetin diglucoside, HDG) and hydroxycinnamic acids (p-couramic, caffeic and ferulic acid glucosides). Their antioxidant and/or cancer chemopreventive properties support their interest in human health and
Gregory M Pask et al.
PloS one, 6(12), e28774-e28774 (2011-12-17)
Insect odorant receptors (ORs) function as odorant-gated ion channels consisting of a conventional, odorant-binding OR and the Orco coreceptor. While Orco can function as a homomeric ion channel, the role(s) of the conventional OR in heteromeric OR complexes has largely
Marco Schlepütz et al.
PloS one, 7(10), e47344-e47344 (2012-10-12)
The peripheral airway innervation of the lower respiratory tract of mammals is not completely functionally characterized. Recently, we have shown in rats that precision-cut lung slices (PCLS) respond to electric field stimulation (EFS) and provide a useful model to study
Lijie Xuan et al.
Plant, cell & environment, 41(12), 2773-2790 (2018-07-08)
Flavonoids are involved in many physiological processes in plants. TRANSPARENT TESTA 4 (TT4) acts at the first step of flavonoid biosynthesis, and the loss of TT4 function causes a lack of flavonoid. Flavonoid deficiency is reportedly the main cause of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service