Skip to Content
Merck
All Photos(1)

Key Documents

83850

Sigma-Aldrich

Ribonucleic acid from Torula utilis

Mr 5000-8000

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€80.40
100 G
€262.00

€80.40


Estimated to ship on06 April 2025



Select a Size

Change View
25 G
€80.40
100 G
€262.00

About This Item

MDL number:
UNSPSC Code:
12352106
NACRES:
NA.51

€80.40


Estimated to ship on06 April 2025


grade

for molecular biology

Quality Level

form

solid

mol wt

Mr 5000-8000

impurities

≤10% water

solubility

0.1 M acetate buffer pH 5.0: soluble 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

RNA from yeast is used in molecular biology laboratories as a carrier during nucleic acid purifications. When used at 10-20 μg/ml, the tRNA acts as a potent co-precipitant that allows for maximal RNA recovery (when working with limited samples).

Application

Ribonucleic acid from Torula utilis was used as a standard in ribonuclease activity assay of certain ribonuclease family members.[1] It was also used as a standard to determine RNA concentration by orcinol assay.[2]
Suitable for use in nucleic acid purifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Debi Ranjan Tripathy et al.
Analytical biochemistry, 437(2), 126-129 (2013-03-19)
The ribonuclease (RNase) activity of ribonucleases has been assayed by observing the change in fluorescence intensity of ethidium bromide on binding with yeast RNA. The binding of EtBr with RNA was monitored via UV-vis and fluorimetric methods. The degradation of
Russel J Lander et al.
Biotechnology and bioengineering, 79(7), 776-784 (2002-09-05)
Preparative-scale purification of plasmid DNA has been attempted by diverse methods, including precipitation with solvents, salts, and detergents and chromatography with ion-exchange, reversed-phase, and size-exclusion columns. Chromatographic methods such as hydrophobic interaction chromatography (HIC), reversed phase chromatography (RPC), and size

Questions

  1. Is this RNA in a double-stranded form?

    1 answer
    1. This is a fairly crude preparation. The product is not tested to determine the ratio of single versus double stranded RNA. It is likely that both forms are present.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service