Skip to Content
Merck

Skip To

83833

Ribonuclease A from bovine pancreas

90%, powder, white, ~80 U/mg

Synonym(s):

Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
EC Number:
232-646-6
EC Number:
MDL number:
Specific activity:
~80 U/mg
Assay:
90%
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

90%

form

powder

specific activity

~80 U/mg

mol wt

~13,700, Mr ~13700

color

white

foreign activity

protease ≤0.001%

shipped in

wet ice

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

General description

RNase A, Ribonuclease A, is an endoribonuclease that cleaves the phosphodiester bonds of single strand RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end (For example pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG). The highest activity is exhibited with single stranded RNA. RNase A is a single chain polypeptide containing 4 disulfide bridges. In contrast to RNase B, it is not a glycoprotein. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase A can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts. RNAse is inhibited in the presence of heavy metal ions. RNase is also inhibited competitively by DNA.

Application

  • RNase A is used to remove RNA from DNA plasmid and genomic DNA preparations and protein samples.
  • RNase A is also used in RNA sequence analysis and protection assays.
  • RNase A has been used as a tool for computer-aided drug design.
  • RNase A supports the analysis of RNA sequences.
  • RNase A hydrolyze RNA contained in protein samples.
  • Purification of DNA is supported by RNase A.

Features and Benefits

Our highly stable Ribonuclease A, RNase A, is suitable for removal of RNA, RNA sequencing, and DNA purification.

Analysis Note

Protein determined by E.

Other Notes

1 U corresponds to the amount of enzyme which hydrolyzes the RNA at a rate constant k = 1 at 25°C and pH 5.0 (Kunitz-units); M. Kunitz, J. Biol. Chem. 164, 563 (1946)
Sales restrictions may apply


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters