Skip to Content
Merck
All Photos(1)

Key Documents

83685

Sigma-Aldrich

Rhodamine B octadecyl ester perchlorate

≥98.0% (TLC), suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H67ClN2O7
CAS Number:
Molecular Weight:
795.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥98.0% (TLC)

form

crystals

mp

43-45 °C (lit.)

solubility

DMF: soluble
DMSO: soluble

fluorescence

λex 554 nm; λem 575 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.CCCCCCCCCCCCCCCCCCOC(=O)c1ccccc1C2=C3C=C\C(C=C3Oc4cc(ccc24)N(CC)CC)=[N+](\CC)CC

InChI

1S/C46H67N2O3.ClHO4/c1-6-11-12-13-14-15-16-17-18-19-20-21-22-23-24-27-34-50-46(49)40-29-26-25-28-39(40)45-41-32-30-37(47(7-2)8-3)35-43(41)51-44-36-38(31-33-42(44)45)48(9-4)10-5;2-1(3,4)5/h25-26,28-33,35-36H,6-24,27,34H2,1-5H3;(H,2,3,4,5)/q+1;/p-1

InChI key

WPBUPFBTCDYKNM-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilic energy transfer acceptor from lipophilic fluoresceins in fluorescence energy transfer assays for cell fusion[1]; Probe for measuring potassium in fiber-optic sensors[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G N Georgiou et al.
FEBS letters, 250(2), 487-492 (1989-07-03)
Fusion of influenza virus with human erythrocytes at pH 5.2 was followed by fluorescence microscopy using a cooled slow-scan CCD camera. The high sensitivity of the CCD permits repetitive digital imaging of the same cells with minimal photobleaching. The experimental
B.P.H. Schaffar et al.
Analyst, 113, 693-693 (1988)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service