Skip to Content
Merck
All Photos(1)

Documents

82658

Sigma-Aldrich

1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt hydrate

suitable for fluorescence, ≥98% (HPLC)

Synonym(s):

Tetrasodium 1,3,6,8-pyrenetetrasulfonate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H6Na4O12S4 · xH2O
Molecular Weight:
610.43 (anhydrous basis)
Beilstein:
3646090
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

crystals

impurities

≤15% water

suitability

suitable for fluorescence

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H10O12S4.4Na.H2O/c17-29(18,19)11-5-13(31(23,24)25)9-3-4-10-14(32(26,27)28)6-12(30(20,21)22)8-2-1-7(11)15(9)16(8)10;;;;;/h1-6H,(H,17,18,19)(H,20,21,22)(H,23,24,25)(H,26,27,28);;;;;1H2/q;4*+1;/p-4

InChI key

LZDOYGMJVXUZGD-UHFFFAOYSA-J

Application

1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt hydrate has been used as a ligand or a crystallisation additive in facilitating protein−protein contacts through cation−π interactions.

Other Notes

Fluorescent probe

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein camouflage: supramolecular anion recognition by ubiquitin
MalloN M, et al.
Chembiochem, 17(8), 774-783 (2016)
N. Kobayashi
Journal of the Chemical Society. Chemical Communications, 918-918 (1988)
E. Koller
Appl. Fluoresc. Technol., 1, 13-13 (1989)
Bruno Mattei et al.
Chemistry and physics of lipids, 202, 28-37 (2016-12-04)
Detergents are widely used to solubilize and separate biomembrane components. It is therefore relevant to study and understand the mechanistic details underlying detergent-lipid interactions using biomimetic systems. Here, we have investigated in detail the process of membrane permeabilization and the

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service