Skip to Content
Merck

82305

TRIS hydrochloride pH 8.5; 0.05M – DTT 0.001M solution

Synonym(s):

Crystallization solution 39/Kit-No 96827

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

pH

8.5±0.2 ( in neat)

density

1.00 g/mL at 20 °C

storage temp.

2-8°C



Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library