Skip to Content
Merck

Skip To

806552

Phosphate Buffered Saline

pH 7.4, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Phosphate buffered saline, PBS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

pH

7.4

General description

Phosphate-buffered saline (PBS) is a buffer solution used in biological research. It is a water-based salt solution containing sodium phosphate, sodium chloride and, in some formulations, it contains potassium chloride and potassium phosphate. The osmolality and ion concentrations of the solutions match those of the human body (isotonic) and are non-toxic to most cells.

Application

Phosphate-buffered saline has been used:
  • as a reagent in the isolation of human endometrial stem cell/stromal cells (hEnSCs) from the endometrium or menstrual blood
  • has also been used to wash cells during immunofluorescence staining
  • as a solvent for the glucose administered orally in the Glucose Tolerance Tests (GTTs)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4474P2272P4244
description

pH 7.4, liquid, sterile-filtered, suitable for cell culture

description

pH 7.4, liquid, sterile-filtered, suitable for cell culture

description

pH 7.2, liquid, sterile-filtered, suitable for cell culture

description

Autoclaved, pH 7.2 (25 °C)

form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

pH

7.4

pH

7.4

pH

7.2

pH

7.2 (25 °C)

impurities

Endotoxin, tested

impurities

Endotoxin, tested

impurities

Endotoxin, tested

impurities

-

sterility

sterile-filtered

sterility

sterile-filtered

sterility

sterile-filtered

sterility

-


Still not finding the right product?

Explore all of our products under Phosphate Buffered Saline


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters