Skip to Content
Merck

79972

7-(Isobutoxycarbonyloxy)-3H-phenoxazin-3-one

suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

O7-(Isobutyloxycarbonyl)resorufin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H15NO5
CAS Number:
Molecular Weight:
313.30
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97.0% (HPLC)

form

powder

fluorescence

λex 500 nm; λem ~593 nm in 0.1 M Tris pH 8.0 (after cleavage by esterase)

suitability

suitable for fluorescence

SMILES string

CC(C)COC(=O)Oc1ccc2N=C3C=CC(=O)C=C3Oc2c1

InChI

1S/C17H15NO5/c1-10(2)9-21-17(20)22-12-4-6-14-16(8-12)23-15-7-11(19)3-5-13(15)18-14/h3-8,10H,9H2,1-2H3

InChI key

RVDFSVVDUVHHKE-UHFFFAOYSA-N

Other Notes

Fluorogenic indicator for cell viability, cell permeant, ~2-times higher sensitivity than calcein-AM; additionally, 80-90 nm more long-wave shift of spectra. Shows esterase activity


wgk

WGK 3

Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library