Skip to Content
Merck

Skip To

79469

D-Arabino-1,4-lactone

≥97.0% (HPLC)

Synonym(s):

D-Arabinonic acid γ-lactone, D-Arabinono-1,4-lactone, D-Arabonic acid-1,4-lactone

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
MDL number:


assay

≥97.0% (HPLC)

form

crystals

optical activity

[α]/D 72.0±3.0°, c = 1 in H2O

color

white

mp

94 - 100  °C ( (201 - 212 °F ))

storage temp.

−20°C

SMILES string

OC[C@H]1OC(=O)[C@@H](O)[C@@H]1O

InChI

1S/C5H8O5/c6-1-2-3(7)4(8)5(9)10-2/h2-4,6-8H,1H2/t2-,3-,4+/m1/s1

InChI key

CUOKHACJLGPRHD-JJYYJPOSSA-N

Application

D-Arabino-1,4-lactone is used as a substrate to identify, differentiate and characterize stereospecific arabino-1,4-lactone oxidase(s)/ arabinonolactone oxidase(s) (LdALO) involved in vitamin C biosynthesis. D-Arabino-1,4-lactone is used as a precursor for the biosynthesis of erythroascorbate.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters