About This Item
Assay
≥80.0% (HPCE)
form
solid
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 436 nm; λem 484 nm±10 nm in PBS, pH 7.4
suitability
suitable for fluorescence
storage temp.
−20°C
SMILES string
CC(CC(C)(C)N1CCCC(NCCOCCOCCOCCN=[N+]=[N-])=O)C(C1=C2)=CC3=C2OC(C(C(OCC)=O)=C3)=O
InChI
1S/C30H43N5O8/c1-5-42-28(37)24-18-22-17-23-21(2)20-30(3,4)35(25(23)19-26(22)43-29(24)38)10-6-7-27(36)32-8-11-39-13-15-41-16-14-40-12-9-33-34-31/h17-19,21H,5-16,20H2,1-4H3,(H,32,36)
InChI key
RSOCGOAHHWBJFE-UHFFFAOYSA-N
General description
Atto 425 is moderately hydrophilic. Its fluorescence can be excited efficiently in the range 405 - 455 nm.
The azide modification is suitable for reactions with alkyne groups (Huisgen reaction - “Click Chemistry“).
find more information here
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service