Skip to Content
Merck
All Photos(1)

Key Documents

77644

Sigma-Aldrich

ASB 14-4

≥97.0% (TLC)

Synonym(s):

4-{N,N-Dimethyl-N-[3-(tetradecanoylamino)propyl]ammonio}butanesulfonate, N,N-Dimethyl-N-[3-[(1-oxotetradecyl)amino]propyl]-4-sulfo1-butanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48N2O4S
CAS Number:
Molecular Weight:
448.70
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

zwitterionic

Assay

≥97.0% (TLC)

mol wt

448.70 g/mol

technique(s)

electrophoresis: suitable

SMILES string

CCCCCCCCCCCCCC(=O)NCCC[N+](C)(C)CCCCS([O-])(=O)=O

InChI

1S/C23H48N2O4S/c1-4-5-6-7-8-9-10-11-12-13-14-18-23(26)24-19-17-21-25(2,3)20-15-16-22-30(27,28)29/h4-22H2,1-3H3,(H-,24,26,27,28,29)

InChI key

DJYNLMBZWBLUKQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William R Tschantz et al.
Protein expression and purification, 57(2), 163-171 (2007-11-21)
OATP2B1 is an important member of the organic anion transporting polypeptides (OATP) family and is implicated in the intestinal and hepatic disposition of endo- and xenobiotics. The purpose of this work was to produce a highly purified protein for use
Christophe Tastet et al.
Proteomics, 3(2), 111-121 (2003-02-26)
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service