Skip to Content
Merck
All Photos(1)

Key Documents

76455

Sigma-Aldrich

(R)-Sodium 2,3-dihydroxyisovalerate hydrate

≥95.0% (CE)

Synonym(s):

(R)-2,3-Dihydroxy-3-methylbutanoic acid sodium salt, D(−)-α,β-Dihydroxyisovaleric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9O4Na · xH2O
Molecular Weight:
156.11 (anhydrous basis)
Beilstein:
1722372
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (CE)

form

powder

optical purity

enantiomeric excess: ≥98.0%

storage temp.

2-8°C

SMILES string

O.[Na+].CC(C)(O)[C@@H](O)C([O-])=O

InChI

1S/C5H10O4.Na.H2O/c1-5(2,9)3(6)4(7)8;;/h3,6,9H,1-2H3,(H,7,8);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

KGRRZNPTTZIUMS-QTNFYWBSSA-M

Biochem/physiol Actions

Important metabolite of branched chain amino acid pathways.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Hwan Park et al.
Biotechnology and bioengineering, 108(4), 934-946 (2011-03-16)
We have previously reported the development of a 100% genetically defined engineered Escherichia coli strain capable of producing L-valine from glucose with a high yield of 0.38 g L-valine per gram glucose (0.58 mol L-valine per mol glucose) by batch culture. Here
Regulatory analysis of amino acid synthesis pathway in Escherichia coli: aspartate family.
Jin, J. H., et al.
Enz. Microbiol. Technol., 35, 694-706 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service