Skip to Content
Merck
All Photos(1)

Key Documents

74781

Sigma-Aldrich

5-Bromo-6-chloro-3-indolyl-β-D-glucopyranoside

≥98.0% (TLC)

Synonym(s):

Magenta-Glc, Magenta glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥98.0% (TLC)

form

powder or crystals

solubility

DMF: 10 mg/mL, clear, colorless to very faint blue

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3cc(Cl)c(Br)cc23)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H15BrClNO6/c15-6-1-5-8(2-7(6)16)17-3-9(5)22-14-13(21)12(20)11(19)10(4-18)23-14/h1-3,10-14,17-21H,4H2/t10-,11-,12+,13-,14-/m1/s1

InChI key

CHRVKCMQIZYLNM-RKQHYHRCSA-N

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

María Moneo-Sánchez et al.
Planta, 251(5), 100-100 (2020-04-25)
β-(1,4)-galactan determines the interactions between different matrix polysaccharides and cellulose during the cessation of cell elongation. Despite recent advances regarding the role of pectic β-(1,4)-galactan neutral side chains in primary cell wall remodelling during growth and cell elongation, little is
Yasunori Suzuki et al.
mSphere, 4(5) (2019-09-27)
The continuous emergence of carbapenemase-producing Enterobacteriaceae (CPE) presents a great public health challenge. Mitigation of CPE spread in the environment is crucial, particularly from a One Health perspective. Here we describe the isolation of CPE strain SNI47 from influent water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service