Skip to Content
Merck
All Photos(1)

Documents

73511

Sigma-Aldrich

4-Nitro-4′-(octadecylamino)stilbene

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

4-(Octadecylamino)-4′-nitrostilbene, N-Octadecyl-4′-nitro-4-stilbenamine, OANS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H48N2O2
CAS Number:
Molecular Weight:
492.74
Beilstein:
4271699
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

mp

130-132 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
toluene: soluble

fluorescence

λex 441 nm; λem ~545 nm in toluene

suitability

suitable for fluorescence

SMILES string

CCCCCCCCCCCCCCCCCCNc1ccc(cc1)\C=C\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C32H48N2O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-28-33-31-24-20-29(21-25-31)18-19-30-22-26-32(27-23-30)34(35)36/h18-27,33H,2-17,28H2,1H3/b19-18+

InChI key

AGCWNVFSUWACRE-VHEBQXMUSA-N

Other Notes

Lipophilic stilbene-dye for use in mixed-dye Langmuir-Blodgett films as non-linear optical material

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.E. Lippitsch et al.
Organic Materials for Non-linear OpticsII, R.A.Hann & D.Bloor eds., 89-89 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service