Skip to Content
Merck
All Photos(1)

Key Documents

73068

Sigma-Aldrich

Magnesium formate solution

BioUltra, 0.5 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2MgO4
Molecular Weight:
114.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

concentration

0.5 M in H2O

impurities

insoluble matter, passes filter test

pH

7.0-7.8 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M, clear, colorless

density

1.038 g/mL at 20 °C

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤500 mg/kg
Bi: ≤1 mg/kg
Ca: ≤500 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤200 mg/kg
Li: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤200 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤50 mg/kg
Zn: ≤1 mg/kg

λ

0.5 M in H2O (20°C)

UV absorption

λ: 260 nm Amax: 0.02
λ: 280 nm Amax: 0.015

SMILES string

[Mg++].[O-]C=O.[O-]C=O

InChI

1S/2CH2O2.Mg/c2*2-1-3;/h2*1H,(H,2,3);/q;;+2/p-2

InChI key

GMDNUWQNDQDBNQ-UHFFFAOYSA-L

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service