Skip to Content
Merck

73034

Polyethylene glycol solution

greener alternative

BioUltra, Molecular Biology, 1,500, ~50% in H2O

Synonym(s):

PEG solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352104
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

product line

BioUltra

form

solution

mol wt

Mr 1400-1600

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

~50% in H2O

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

pH

6.0-7.5 (25 °C)

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

UV absorption

λ: 260 nm Amax: 0.9, λ: 280 nm Amax: 0.3

greener alternative category

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.

Application

PEG is used as a fusogen to obtain hybridomas for monoclonal antibody production. Induces cell hybridization.


Still not finding the right product?

Explore all of our products under Polyethylene glycol solution


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library