71790
Sodium nucleinate
Synonym(s):
Ribonucleic acid sodium salt
Select a Size
About This Item
Recommended Products
form
powder
loss
~5% loss on drying, 20 °C (HV)
solubility
H2O: 0.1 g/mL, clear to turbid, brown-yellow (hot)
storage temp.
2-8°C
General description
Application
<li><strong>Tetrahydrophthalazine derivative "sodium nucleinate" exert its anti-inflammatory effects through inhibition of oxidative burst in human monocytes:</strong> Describes the anti-inflammatory capabilities of Sodium nucleinate, particularly its ability to inhibit oxidative burst in human monocytes, suggesting its use in anti-inflammatory therapies (Jukic et al., 2012).</li>
<li><strong>A tetrahydrophthalazine derivative ′sodium nucleinate′ exerts a potent suppressive effect upon LPS-stimulated mononuclear cells in vitro and in vivo:</strong> Outlines the suppressive effects of Sodium nucleinate on LPS-stimulated mononuclear cells, underlining its potential utility in controlling inflammatory responses (Jukic et al., 2011).</li>
</ul>
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service