Skip to Content
Merck

Skip To

69234

Mant-GTP triethylammonium salt solution

≥95.0% (HPLC)

Synonym(s):

2′/3′-O-(N-Methylanthraniloyl)guanosine 5′-triphosphate triethylammonium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 μmol
Check Cart for Availability
€ 208,00

About This Item

Empirical Formula (Hill Notation):
C18H23N6O15P3 · xC6H15N
CAS Number:
Molecular Weight:
656.33 (free acid basis)
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32
Assay:
≥95.0% (HPLC)
Form:
liquid
Storage temp.:
−20°C

€ 208,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95.0% (HPLC)

form

liquid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(OP(O)(OP(O)(OC[C@@H]1[C@@H]([R1])[C@@H]([R2])[C@H](N(C=N2)C3=C2C(NC(N)=N3)=O)O1)=O)=O)(O)=O.CCN(CC)CC.O=C(C4=C(NC)C=CC=C4)O.O=C(C5=C(NC)C=CC=C5)O


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
187236924419511
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

assay

≥95.0% (HPLC)

form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

pH

7.0-8.0

pH

7.0-8.0

pH

7.0-8.0

pH

7.0-8.0


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters