Skip to Content
Merck

Skip To

67455

Atto 514 NHS ester

suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108


assay

90% (Coupling efficiency with benzylamine, HPLC)

form

solid

mol wt

Mw 1111 g/mol

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm, 514 nm

fluorescence

λex 511 nm; λem 533 nm±10 nm in PBS, pH 7.4

λ

(water/methanol with 0.1% perchloric acid)

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 514 is a new hydrophilic fluorescent label with excellent water solubility. The dye exhibits strong absorption, high fluorescence quantum yield and exceptional thermal and photo-stability. Thus Atto 514 is highly suitable for single-molecule detection applications and high-resolution microscopy such as PALM, dSTORM, STED etc. Additionally the dye highly qualifies to be applied in flow cytometry (FACS), fluorescence in-situ hybridization (FISH) and many more. The fluorescence is excited most efficiently in the range 510 - 535 nm. A suitable source of excitation is the 514 nm line of the Argon-Ion laser.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
525957836268606
suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

-

fluorescence

λex 511 nm; λem 533 nm±10 nm in PBS, pH 7.4

fluorescence

λex 511 nm; λem 533 nm±10 nm in PBS, pH 7.4

fluorescence

-

fluorescence

-

form

solid

form

solid

form

solid

form

solid

assay

90% (Coupling efficiency with benzylamine, HPLC)

assay

≥90.0% (HPLC)

assay

≥90% (HPLC), ≥90% (degree of coupling)

assay

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

transmittance

254 nm, 514 nm

transmittance

254 nm, 514 nm

transmittance

-

transmittance

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters