Skip to Content
Merck

Skip To

64951

7-Methoxycoumarin

suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

Herniarin, Methyl umbelliferyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 214,00
€ 149,80
500 mg
Check Cart for Availability
€ 776,00

About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
UNSPSC Code:
85151701
NACRES:
NA.32
PubChem Substance ID:
EC Number:
208-513-3
Beilstein/REAXYS Number:
141728
MDL number:

€ 149,80

List Price€ 214,00Save 30%
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (TLC)

form

solid

mp

117-121 °C (lit.)

solubility

DMF: soluble, acetonitrile: soluble, alcohols: soluble, chloroform: soluble

fluorescence

λex 350 nm; λem 385 nm (Reaction product), λex 350 nm; λem 385 nm in chloroform

suitability

suitable for fluorescence

SMILES string

COc1ccc2C=CC(=O)Oc2c1

InChI

1S/C10H8O3/c1-12-8-4-2-7-3-5-10(11)13-9(7)6-8/h2-6H,1H3

InChI key

LIIALPBMIOVAHH-UHFFFAOYSA-N

Application

7-Methoxycoumarin (Herniarin) is used to study antioxidant and hepatoprotective properties and its activity as an allergen. 7-Methoxycoumarin may be used as a reference material in the purification, separation and analysis of coumarin compounds.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3165PHL89733T2803
assay

≥98.0% (TLC)

assay

≥99% (TLC)

assay

≥98.0% (HPLC)

assay

≥98% (TLC)

mp

117-121 °C (lit.)

mp

-

mp

117-121 °C (lit.)

mp

-

form

solid

form

powder

form

crystals

form

powder

fluorescence

λex 350 nm; λem 385 nm (Reaction product), λex 350 nm; λem 385 nm in chloroform

fluorescence

λex 333 nm; λem 416 nm in methanol

fluorescence

-

fluorescence

λex 333 nm; λem 415 nm in methanol

suitability

suitable for fluorescence

suitability

-

suitability

-

suitability

-

Quality Level

100

Quality Level

200

Quality Level

-

Quality Level

100


Still not finding the right product?

Explore all of our products under 7-Methoxycoumarin


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters