Skip to Content
Merck

Skip To

64948

7-Methoxycoumarin-3-carboxylic acid

suitable for fluorescence, ≥97.0% (HPCE)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C11H8O5
CAS Number:
Molecular Weight:
220.18
UNSPSC Code:
12352200
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
384199


assay

≥97.0% (HPCE)

form

solid

solubility

DMF: soluble, DMSO: soluble, acetonitrile: soluble

fluorescence

λex 330 nm; λem 402 nm in 0.1 M Tris pH 9.0

suitability

suitable for fluorescence

SMILES string

COc1ccc2C=C(C(O)=O)C(=O)Oc2c1

InChI

1S/C11H8O5/c1-15-7-3-2-6-4-8(10(12)13)11(14)16-9(6)5-7/h2-5H,1H3,(H,12,13)

InChI key

VEEGNDSSWAOLFN-UHFFFAOYSA-N

Application

7-Methoxycoumarin-3-carboxylic acid N-succinimidyl ester is used as a reagent to conjugate 7-Methoxycoumarin-3-carboxylic acid to other molecules via amide chemistry.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters