Skip to Content
Merck
All Photos(4)

Documents

63020-016

Sigma-Aldrich

DNAgard® Tissue

Trial kit, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

for molecular biology

form

liquid

usage

3 reactions

storage temp.

room temp

Features and Benefits

  • Protects DNA in tissues, cells and biopsy material from degradation at room temperature
  • Compatible with most downstream applications, e.g., PCR, qPCR and sequencing.
  • Optional dry-down feature allows for long-term storage
  • Reduces storage and shipping costs and improves lab space management

Packaging

3 tubes x 500 μL

Legal Information

DNAgard is a registered trademark of Biomatrica

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven B Lee et al.
Forensic science international. Genetics, 6(1), 31-40 (2011-02-18)
The ability to properly collect, analyze and preserve biological stains is important to preserving the integrity of forensic evidence. Stabilization of intact biological evidence in cells and the DNA extracts from them is particularly important since testing is generally not
Michael A Gray et al.
FEMS microbiology ecology, 83(2), 468-477 (2012-09-15)
Field collections of environmental samples, for example corals, for molecular microbial analyses present distinct challenges. The lack of laboratory facilities in remote locations is common, and preservation of microbial community DNA for later study is critical. A particular challenge is
A Allen-Hall et al.
Forensic science international. Genetics, 6(5), 653-657 (2012-01-25)
Disaster victim identification (DVI) poses unique challenges for forensic personnel. Typical scenarios may involve many bodies or body parts to identify in remote locations with limited access to laboratory facilities and in extreme temperatures. Transportation of tissue samples to a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service