Skip to Content
Merck
All Photos(1)

Key Documents

62028

Sigma-Aldrich

6′-Amino-D-luciferin

≥95.0% (HPLC)

Synonym(s):

(S)-2-(6-Amino-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid, (S)-2-(6-Amino-2-benzothiazolyl)-4,5-dihydro-4-thiazolecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€485.00

€485.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€485.00

About This Item

Empirical Formula (Hill Notation):
C11H9N3O2S2
CAS Number:
Molecular Weight:
279.34
Beilstein:
12930905
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

€485.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥95.0% (HPLC)

fluorescence

λex 350 nm; λem 516 nm±10 nm in H2O

storage temp.

−20°C

SMILES string

OC([C@H]1CSC(C(S2)=NC3=C2C=C(N)C=C3)=N1)=O

InChI

1S/C11H9N3O2S2/c12-5-1-2-6-8(3-5)18-10(13-6)9-14-7(4-17-9)11(15)16/h1-3,7H,4,12H2,(H,15,16)/t7-/m1/s1

InChI key

HKSJKXOOBAVPKR-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

596.5 °F

Flash Point(C)

313.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service