Skip to Content
Merck

Skip To

59893

Sodium N-(4-azido-2-nitrophenyl)-2-aminoethylsulfonate dihydrate

≥90% (TLC)

Synonym(s):

2-[(4-Azido-2-nitrophenyl)amino]ethanesulfonic acid monosodium salt dihydrate, N-(4-Azido-2-nitrophenyl) taurine sodium salt dihydrate, N-(4-Azido-2-nitrophenyl)-2-aminoethylsulfonic acid sodium salt dihydrate, NAP-Taurine sodium salt dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 1.630,00

About This Item

Empirical Formula (Hill Notation):
C8H8N5NaO5S · 2H2O
CAS Number:
Molecular Weight:
345.26
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

€ 1.630,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% (TLC)

form

solid

storage temp.

−20°C

SMILES string

O.O.[Na+].[O-][N+](=O)c1cc(ccc1NCCS([O-])(=O)=O)N=[N+]=[N-]

InChI

1S/C8H9N5O5S.Na.2H2O/c9-12-11-6-1-2-7(8(5-6)13(14)15)10-3-4-19(16,17)18;;;/h1-2,5,10H,3-4H2,(H,16,17,18);;2*1H2/q;+1;;/p-1

InChI key

AQXKSSHRIZHNEM-UHFFFAOYSA-M

General description

brick-red powder; soluble in water and Ethanol; labeling reagent (Photolysis of NAP-taurine produces a highly reactive nitrene which acts as a general covalent labeling reagent)

Analysis Note

contains up to 10.4% water (dihydrate)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
228729719951614909
Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

-

assay

≥90% (TLC)

assay

≥99%

assay

≥98.0% (NT)

assay

-

storage temp.

−20°C

storage temp.

-

storage temp.

-

storage temp.

-

form

solid

form

-

form

solid

form

-


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters