Skip to Content
Merck
All Photos(1)

Key Documents

58756

Sigma-Aldrich

1-(5-Isoquinolinesulfonyl)-3-methylpiperazine

≥98.0% (HPLC)

Synonym(s):

1-(5-Isoquinolinylsulfonyl)-3-methylpiperazine, Iso-H-7

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30

€89.30


Estimated to ship on22 May 2025



Select a Size

Change View
5 MG
€89.30

About This Item

Empirical Formula (Hill Notation):
C14H17N3O2S
CAS Number:
Molecular Weight:
291.37
Beilstein:
5445506
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€89.30


Estimated to ship on22 May 2025


Assay

≥98.0% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

CC1CN(CCN1)S(=O)(=O)c2cccc3cnccc23

InChI

1S/C14H17N3O2S/c1-11-10-17(8-7-16-11)20(18,19)14-4-2-3-12-9-15-6-5-13(12)14/h2-6,9,11,16H,7-8,10H2,1H3

InChI key

QGSCXAWTTISKLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Less potent inhibitor of protein kinaseC than H-7

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D Balaban et al.
Brain research, 845(1), 97-101 (1999-10-26)
This study tests the hypothesis that activation of protein kinase C (PKC) is a critical step for early recovery from spontaneous nystagmus after unilateral ablation of the vestibular periphery. Halothane-NO(2)-O(2)-anesthetized Long-Evans rats received a 5-microl intracerebroventricular bolus of vehicle (distilled
C A Gebert et al.
Molecular endocrinology (Baltimore, Md.), 13(1), 38-56 (1999-01-19)
STAT5b (signal transducer and activator of transcription 5b) is a key mediator of the effects of plasma GH pulses on male-specific liver gene expression. STAT5b is activated in liver cells in vivo by physiological pulses of GH and then is
L B Quesnel et al.
The Journal of antimicrobial chemotherapy, 41(2), 215-221 (1998-04-09)
Cystic fibrosis (CF) results from mutations in the gene encoding the CF transmembrane conductance regulator (CFTR) which is a regulated chloride channel. The deltaF508 mutation prevents the post-translational glycosylation and membrane insertion of the protein. Severe disease follows, with the
A Broberg et al.
Biochemical and biophysical research communications, 280(1), 328-333 (2001-02-13)
Integrin alpha1beta1, one of the cellular collagen receptors, can participate in the regulation of collagen accumulation by acting as a negative feedback regulator. The molecular mechanism behind this phenomenon has been unknown. We have plated cells inside three-dimensional collagen and
K Goishi et al.
Molecular biology of the cell, 6(8), 967-980 (1995-08-01)
Vero cell heparin-binding epidermal growth factor-like growth factor (HB-EGF) is synthesized as a 20- to 30-kDa membrane-anchored HB-EGF precursor (proHB-EGF). Localization and processing of proHB-EGF, both constitutive and 12-O-tetradecanoylphorbol 13-acetate (TPA)-inducible, was examined in Vero cells overexpressing recombinant HB-EGF (Vero

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service