Skip to Content
Merck
All Photos(1)

Key Documents

56962

Sigma-Aldrich

D-myo-Inositol 1,2,4,5,6-pentakis-phosphate

≥96% (HPLC)

Synonym(s):

L-myo-Inositol 2,3,4,5,6-pentakis-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17O21P5
CAS Number:
Molecular Weight:
580.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥96% (HPLC)

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O

InChI

1S/C6H17O21P5/c7-1-2(23-28(8,9)10)4(25-30(14,15)16)6(27-32(20,21)22)5(26-31(17,18)19)3(1)24-29(11,12)13/h1-7H,(H2,8,9,10)(H2,11,12,13)(H2,14,15,16)(H2,17,18,19)(H2,20,21,22)/t1-,2-,3-,4+,5-,6-/m1/s1

InChI key

CTPQAXVNYGZUAJ-XCMZKKERSA-N

Biochem/physiol Actions

Important intermediate in inositol phosphate metabolism. Inositides have an important impact on diverse areas of cellular regulation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean G Jackson et al.
BMC structural biology, 11, 11-11 (2011-02-12)
PH domains represent one of the most common domains in the human proteome. These domains are recognized as important mediators of protein-phosphoinositide and protein-protein interactions. Phosphoinositides are lipid components of the membrane that function as signaling molecules by targeting proteins
Nathalie Hayez et al.
Journal of neuroimmunology, 149(1-2), 167-181 (2004-03-17)
The neuropeptides Vasoactive-intestinal peptide (VIP) and Pituitary adenylate-cyclase activating protein (PACAP) increased cAMP levels in three out of five human myeloid leukemic cell lines tested while an increased in calcium intracytoplasmic levels was seen only in one cell line (HEL).
Laura B Sheard et al.
Nature, 468(7322), 400-405 (2010-10-12)
Jasmonates are a family of plant hormones that regulate plant growth, development and responses to stress. The F-box protein CORONATINE INSENSITIVE 1 (COI1) mediates jasmonate signalling by promoting hormone-dependent ubiquitylation and degradation of transcriptional repressor JAZ proteins. Despite its importance
Ralf Greiner
The protein journal, 23(8), 577-585 (2005-01-15)
High-pressure liquid chromatography (HPLC) analysis established myo-inositol pentakisphosphate as the final product of phytate dephosphorylation by the phytate-degrading enzyme from Pantoea agglomerans. Neither product inhibition by phosphate nor inactivation of the Pantoea enzyme during the incubation period were responsible for
Samuel E K Caddick et al.
FEBS letters, 582(12), 1731-1737 (2008-04-30)
We describe a multifunctional inositol polyphosphate kinase/phosphotransferase from Solanum tuberosum, StITPKalpha (GenBank accession number: EF362784), hereafter called StITPK1. StITPK1 displays inositol 3,4,5,6-tetrakisphosphate 1-kinase activity: K(m) = 27 microM, and V(max) = 19 nmol min(-1) mg(-1). The enzyme displays inositol 1,3,4,5,6-pentakisphosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service