Skip to Content
Merck
All Photos(1)

Key Documents

56064C

SAFC

Dulbecco′s Phosphate Buffered Saline

dry powder, DPBS Modified, without calcium, without magnesium, suitable for cell culture

Pharma Manufacturing

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing

Select a Size

10 L
€27.00
50 L
€88.00

€27.00


Estimated to ship on16 April 2025



Select a Size

Change View
10 L
€27.00
50 L
€88.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.71

€27.00


Estimated to ship on16 April 2025


Quality Level

description

for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Phosphate Buffered Saline (DPBS) is intended for use in the maintenance of mammalian cells where a chemically defined balanced salt solution provides an environment that will maintain the structural and physiological integrity of cells in vitro. DPBS is commonly used in cell enumerations as a diluent, for rinsing cells and as a buffer in many chromatographic procedures. DPBS also is used to wash and resuspend cells during the dissociation process, where the presence of calcium and magnesium can inhibit trypsin activity. For this application, use DPBS Modified, which is formulated without calcium and magnesium salts.
Dulbecco′s phosphate buffered saline (DPBS) has been used in in vitro degradation studies of specimens.[1]

Quantity

Formulated to contain 9.6 grams of powder per liter of medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of curing regime and maturation time on photopolymerisation and in vitro behavior of a polymeric light-cured calcium phosphate cement.
Barounian M H, et al.
Biointerface Research in Applied Chemistry, 6(3) (2016)
Fahimeh Shahabipour et al.
Journal of biomedical materials research. Part A, 108(8), 1596-1606 (2020-03-18)
The interaction between osteogenic and angiogenic cells through a coculturing system in biocompatible materials has been considered for successfully engineering vascularized bone tissue equivalents. In this study, we developed a hydrogel-blended scaffold consisted of gelatin methacryloyl (GelMA) and alginate enriched

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service