Skip to Content
Merck
All Photos(3)

Documents

53201-013

Sigma-Aldrich

RNAstable® LD

greener alternative

2 ml, for room temp preservation of RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

grade

for molecular biology

form

liquid

usage

100 reactions

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

RNAstable® preserves RNA samples at room temperature. Each tube or plate contains RNAstable® as a coating at the bottom of the tube or well, which protects picogram to microgram amounts of RNA. This medium is completely dissolvable and ensures total and quantitative sample recovery.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Features and Benefits


  • Preserves RNA at ambient temperatures without degradation
  • Eliminates the need to store RNA in freezers or to ship on dry ice
  • Concentrates RNA more effectively than other methods
  • Compatible with most downstream applications

Packaging

  • (1) RNAstable® LD Tube
  • Quick Reference Protocol
  • 2 Desiccant packets
  • Re-sealable moisture-barrier foil pouch

Legal Information

Biomatrica is a registered trademark of Biomatrica
RNAstable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonggang Zhang et al.
BMC medical genomics, 11(1), 78-78 (2018-09-12)
CRISPR/CAS9 (epi)genome editing revolutionized the field of gene and cell therapy. Our previous study demonstrated that a rapid and robust reactivation of the HIV latent reservoir by a catalytically-deficient Cas9 (dCas9)-synergistic activation mediator (SAM) via HIV long terminal repeat (LTR)-specific
Adrian Puren et al.
The Journal of infectious diseases, 201 Suppl 1, S27-S36 (2010-03-17)
RNA remains the most informative and accurate biomarker for human immunodeficiency virus type 1 load diagnostics and for surveillance of drug resistance markers. Viral load testing by nucleic acid amplification currently is a complex and expensive test that is restricted
Daniel S Stevens et al.
Journal of virological methods, 182(1-2), 104-110 (2012-03-22)
Two approaches to stabilize viral nucleic acid in processed clinical specimens were evaluated. HIV-1 RNA extracted from clinical specimens was stabilized in a dry matrix in a commercial product (RNAstable, Biomatrica, San Diego, CA, USA) and in a reverse-transcription reaction
Gilberto E Hernandez et al.
BioTechniques, 47(2), 667-668 (2009-09-10)
RNA integrity is a critical factor in obtaining meaningful gene expression data. Current methodologies rely on maintaining samples in cold environments during collection, transport, processing, and storage procedures, which are also extremely time-sensitive. Several RNA storage products are commercially available
Wen Huang et al.
PloS one, 7(1), e30244-e30244 (2012-02-01)
In this study, mRNA-Seq was used to characterize and compare the leukocyte transcriptomes from two taurine breeds (Holstein and Jersey), and one indicine breed (Cholistani). At the genomic level, we identified breed-specific base changes in protein coding regions. Among 7,793,425

Articles

RNAstable® preserves RNA via anhydrobiosis principles, enabling ambient temp storage and shipping.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service