Skip to Content
Merck

Skip To

51410

Nε-(N-(+)-Biotinyl-6-aminohexanoyl)-Nα,Nα-bis(carboxymethyl)-L-lysine tripotassium salt

≥98.0% (TLC)

Synonym(s):

Biotin-X-NTA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 284,00

About This Item

Empirical Formula (Hill Notation):
C26H40K3N5O9S
CAS Number:
Molecular Weight:
715.98
UNSPSC Code:
12352209
NACRES:
NA.32
Assay:
≥98.0% (TLC)
Form:
solid

€ 284,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (TLC)

form

solid

SMILES string

[K+].[K+].[K+].[O-]C(=O)CN(CC([O-])=O)[C@H](CCCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)C([O-])=O

InChI

1S/C26H43N5O9S.3K/c32-20(28-13-7-5-8-18(25(38)39)31(14-22(34)35)15-23(36)37)10-2-1-6-12-27-21(33)11-4-3-9-19-24-17(16-41-19)29-26(40)30-24;;;/h17-19,24H,1-16H2,(H,27,33)(H,28,32)(H,34,35)(H,36,37)(H,38,39)(H2,29,30,40);;;/q;3*+1/p-3/t17-,18+,19-,24-;;;/m0

InChI key

MKLSOXPHWHEJPK-NERIWEOLSA-K

Application

Nε-(N-(+)-Biotinyl-6-aminohexanoyl)-Nα,Nα-bis(carboxymethyl)-L-lysine (Biotin-X-NTA) is used as a hetero-bifunctional linker between histone(His)-tagged proteins and (strept)avidin conjugates.
Hetero-bifunctional linker for His-tagged proteins on one side and (strept)avidin conjugates[1][2][3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
737491440775543
form

solid

form

powder

form

powder

form

liquid

assay

≥98.0% (TLC)

assay

≥95.0% (HPLC)

assay

≥97.0% (HPLC)

assay

≥80.0% (HPLC)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters